Gene Transcripts

Gene Symbol
OR51B2
Entrez Gene ID
79345
Gene Name
olfactory receptor family 51 subfamily B member 2 (gene/pseudogene)
Gene Aliases
HOR5'Beta3, OR51B1P
Chromosome Location
Chr. 11: 5323298 - 5324352 on Build GRCh38
UniGene ID
Hs.680163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033180.4 NP_149420.4 1 486
GenBank mRNA BC117461.1 - 1 486

View Full Product Details