Gene Transcripts

Gene Symbol
FAM71A
Entrez Gene ID
149647
Gene Name
family with sequence similarity 71 member A
Gene Aliases
Chromosome Location
Chr. 1: 212624447 - 212626778 on Build GRCh38
UniGene ID
Hs.129293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153606.3 NP_705834.2 1 1055
GenBank mRNA AK057358.1 - 1 668
AK315021.1 - 1 679
BC035007.1 - 1 1030 5267155

View Full Product Details