Gene Transcripts
- Gene Symbol
- WFDC13
- Entrez Gene ID
- 164237
- Gene Name
- WAP four-disulfide core domain 13
- Gene Aliases
- C20orf138, WAP13, dJ601O1.3
- Chromosome Location
- Chr. 20: 45702016 - 45708817 on Build GRCh38
- UniGene ID
- Hs.406733
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172005.1 |
NP_742002.1
|
1, 2 |
196 |
|
GenBank mRNA |
AF454505.1 |
- |
1, 2 |
196 |
|
View Full Product Details