Gene Transcripts

Gene Symbol
OR7E24
Entrez Gene ID
26648
Gene Name
olfactory receptor family 7 subfamily E member 24
Gene Aliases
HSHT2, OR19-8, OR7E24P, OR7E24Q
Chromosome Location
Chr. 19: 9251044 - 9252063 on Build GRCh38
UniGene ID
Hs.129832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079935.1 NP_001073404.1 1 326
GenBank mRNA BC146920.1 - 1 440
Y10529.1 - 1 101

View Full Product Details