Gene Transcripts

Gene Symbol
HIGD1C
Entrez Gene ID
613227
Gene Name
HIG1 hypoxia inducible domain family member 1C
Gene Aliases
GM921
Chromosome Location
Chr. 12: 50936223 - 50972592 on Build GRCh38
UniGene ID
Hs.702849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109619.1 NP_001103089.1 3 261
XM_017019784.1 XP_016875273.1 6 1836

View Full Product Details