Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GMPPA
Entrez Gene ID
29926
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
AAMR
Chromosome Location
Chr.2: 219498865 - 219506996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013335.3 NP_037467.2 7 - 8 718 61
NM_205847.2 NP_995319.1 7 - 8 999 61
XM_005246486.3 XP_005246543.1 7 - 8 687 61
XM_017003909.1 XP_016859398.1 7 - 8 718 61
XM_017003910.1 XP_016859399.1 7 - 8 687 61
XM_017003911.1 XP_016859400.1 7 - 8 993 61
XM_017003916.1 XP_016859405.1 7 - 8 647 61
XM_017003917.1 XP_016859406.1 7 - 8 647 61
GenBank mRNA AF135422.2 - 6 - 7 697 61
AK000999.1 - 7 - 8 692 61
AK022578.1 - 7 - 8 993 61
AK222951.1 - 7 - 8 682 61
AK290671.1 - 7 - 8 995 61
BC007456.2 - 7 - 8 987 61

View Full Product Details