The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014448.3 | NP_055263.2 | 2 - 3 | 837 | 72 |
XM_017001049.1 | XP_016856538.1 | 2 - 3 | 1438 | 72 | |
XM_017001050.1 | XP_016856539.1 | 2 - 3 | 643 | 72 | |
XM_017001051.1 | XP_016856540.1 | 2 - 3 | 848 | 72 | |
GenBank mRNA | AK095984.1 | - | 2 - 3 | 744 | 72 |
BC002681.1 | - | 1 - 2 | 124 | 72 |