The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014503.2 | NP_055318.2 | 29 - 30 | 3802 | 83 |
GenBank mRNA | AJ006778.1 | - | 29 - 30 | 3790 | 83 |