Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC51
Entrez Gene ID
79714
Gene Name
coiled-coil domain containing 51
Gene Aliases
-
Chromosome Location
Chr.3: 48432170 - 48446652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256964.1 NP_001243893.1 2 - 3 453 55
NM_001256966.2 NP_001243895.1 2 - 3 332 55
NM_001256967.2 NP_001243896.1 2 - 3 327 55
NM_001256968.2 NP_001243897.1 2 - 3 224 55
NM_001256969.2 NP_001243898.1 2 - 3 219 55
NM_024661.4 NP_078937.3 2 - 3 345 55
XM_011534113.2 XP_011532415.1 3 - 4 780 55
GenBank mRNA AK022498.1 - 2 - 3 224 55
AW957929.1 - 2 - 3 229 55
BC011993.1 - 1 - 2 335 55
BM810621.1 - 2 - 3 355 55
BP307614.1 - 2 - 3 345 55
BQ050243.1 - 2 - 3 267 55
BQ427785.1 - 1 - 2 229 55

View Full Product Details