The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177965.3 | NP_808880.1 | 1 - 2 | 204 | 101 |
XM_005250799.3 | XP_005250856.2 | 1 - 2 | 1572 | 101 | |
GenBank mRNA | AK055162.1 | - | 1 - 2 | 179 | 101 |
BC036855.2 | - | 1 - 2 | 169 | 101 | |
DA473498.1 | - | 1 - 2 | 204 | 101 | |
DB205618.1 | - | 1 - 2 | 169 | 101 | |
GQ240139.1 | - | 1 - 2 | 179 | 101 |