The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323916.1 | NP_001310845.1 | 17 - 17 | 5412 | 101 |
NM_020840.2 | NP_065891.1 | 17 - 17 | 5407 | 101 | |
XM_005263156.1 | XP_005263213.1 | 18 - 18 | 5497 | 101 | |
XM_005263158.1 | XP_005263215.1 | 18 - 18 | 5297 | 101 | |
XM_011532154.2 | XP_011530456.1 | 18 - 18 | 6277 | 101 | |
XM_017008487.1 | XP_016863976.1 | 19 - 19 | 5446 | 101 | |
GenBank mRNA | AK057981.1 | - | 1 - 1 | 353 | 101 |
BC047069.1 | - | 1 - 1 | 1247 | 101 | |
BU753084.1 | - | 1 - 1 | 101 |