The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138294.2 | NP_612151.1 | 4 - 5 | 255 | 61 |
GenBank mRNA | AF462605.1 | - | 4 - 5 | 251 | 61 |
BC039863.1 | - | 4 - 5 | 255 | 61 | |
BC107044.2 | - | 3 - 4 | 215 | 61 |