The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015124.4 | NP_055939.1 | 9 - 10 | 1019 | 93 |
XM_005261396.3 | XP_005261453.1 | 9 - 10 | 955 | 93 | |
XM_006724169.1 | XP_006724232.1 | 9 - 10 | 917 | 93 | |
XM_006724170.3 | XP_006724233.1 | 9 - 10 | 936 | 93 | |
XM_011530011.1 | XP_011528313.1 | 8 - 9 | 855 | 93 | |
XM_011530012.1 | XP_011528314.1 | 8 - 9 | 855 | 93 | |
XM_011530013.2 | XP_011528315.2 | 9 - 10 | 964 | 93 | |
XM_011530014.2 | XP_011528316.1 | 8 - 9 | 855 | 93 | |
XM_017028670.1 | XP_016884159.1 | 9 - 10 | 977 | 93 | |
GenBank mRNA | AB018310.2 | - | 9 - 10 | 1019 | 93 |
BC129837.1 | - | 8 - 9 | 840 | 93 | |
CR456470.1 | - | 8 - 9 | 845 | 93 |