Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMND5B
Entrez Gene ID
64777
Gene Name
required for meiotic nuclear division 5 homolog B
Gene Aliases
GID2, GID2B
Chromosome Location
Chr.5: 178130532 - 178148570 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288794.1 NP_001275723.1 8 - 9 1083 62
NM_001288795.1 NP_001275724.1 7 - 8 932 62
NM_022762.4 NP_073599.2 7 - 8 943 62
XM_005265969.3 XP_005266026.1 6 - 7 763 62
XM_005265971.4 XP_005266028.1 7 - 8 1510 62
XM_005265972.3 XP_005266029.1 6 - 7 890 62
XM_017009727.1 XP_016865216.1 6 - 7 1554 62
XM_017009728.1 XP_016865217.1 6 - 7 1221 62
GenBank mRNA AK025971.1 - 4 - 5 488 62
AK093481.1 - 7 - 8 884 62
AK094065.1 - 4 - 5 1090 62
AK122692.1 - 2 - 3 1806 62
AK125337.1 - 8 - 9 1030 62
AL137381.1 - 1 - 2 56 62
AL713670.1 - 6 - 7 806 62
AY359092.1 - 4 - 5 1801 62
BC009911.2 - 7 - 8 877 62
DQ494789.1 - 7 - 8 877 62

View Full Product Details