Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP10-10
Entrez Gene ID
353333
Gene Name
keratin associated protein 10-10
Gene Aliases
KAP10.10, KAP18.10, KRTAP18-10, KRTAP18.10
Chromosome Location
Chr.21: 44637356 - 44638455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181688.2 NP_859016.1 1 - 1 840 80
GenBank mRNA AB076358.1 - 1 - 1 825 80
AJ566387.1 - 1 - 1 840 80

Gene Symbol
KRTAP10-6
Entrez Gene ID
386674
Gene Name
keratin associated protein 10-6
Gene Aliases
KAP10.6, KAP18.6, KRTAP18-6, KRTAP18.6
Chromosome Location
Chr.21: 44591268 - 44592505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198688.3 NP_941961.3 1 - 1 1141 80
GenBank mRNA AB076353.1 - 1 - 1 1141 80
BC140725.1 - - 1154 80

View Full Product Details