The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197259.1 | NP_001184188.1 | 3 - 4 | 323 | 119 |
NM_012381.3 | NP_036513.2 | 4 - 5 | 421 | 119 | |
NM_181837.2 | NP_862820.1 | 4 - 5 | 421 | 119 | |
XM_005248704.1 | XP_005248761.1 | 4 - 5 | 367 | 119 | |
XM_005248705.1 | XP_005248762.1 | 5 - 6 | 425 | 119 | |
XM_005248706.1 | XP_005248763.1 | 5 - 6 | 424 | 119 | |
XM_011535651.1 | XP_011533953.1 | 4 - 5 | 367 | 119 | |
XM_011535652.2 | XP_011533954.1 | 4 - 5 | 367 | 119 | |
XM_017010632.1 | XP_016866121.1 | 4 - 5 | 367 | 119 | |
XM_017010633.1 | XP_016866122.1 | 4 - 5 | 367 | 119 | |
XM_017010634.1 | XP_016866123.1 | 4 - 5 | 367 | 119 | |
XM_017010635.1 | XP_016866124.1 | 4 - 5 | 367 | 119 | |
XM_017010636.1 | XP_016866125.1 | 5 - 6 | 425 | 119 | |
XM_017010637.1 | XP_016866126.1 | 5 - 6 | 424 | 119 | |
XM_017010638.1 | XP_016866127.1 | 5 - 6 | 425 | 119 | |
XM_017010639.1 | XP_016866128.1 | 5 - 6 | 424 | 119 | |
XM_017010640.1 | XP_016866129.1 | 5 - 6 | 424 | 119 | |
GenBank mRNA | AF093535.1 | - | 4 - 5 | 345 | 119 |
AF125507.1 | - | 4 - 5 | 354 | 119 | |
AF135044.1 | - | 4 - 5 | 319 | 119 | |
AK222795.1 | - | 4 - 5 | 347 | 119 | |
AK302750.1 | - | 5 - 6 | 415 | 119 | |
AK303056.1 | - | 4 - 5 | 347 | 119 | |
AK303195.1 | - | 3 - 4 | 247 | 119 | |
AK315911.1 | - | 4 - 5 | 308 | 119 | |
AK316298.1 | - | 4 - 5 | 352 | 119 | |
AL080116.1 | - | 4 - 5 | 360 | 119 | |
BC035494.2 | - | 4 - 5 | 336 | 119 | |
BC047689.1 | - | 4 - 5 | 337 | 119 | |
BF215571.1 | - | - | 363 | 119 | |
DC395106.1 | - | 4 - 5 | 421 | 119 |