The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145444.1 | NP_001138916.1 | 3 - 4 | 695 | 140 |
NM_001145445.1 | NP_001138917.1 | 4 - 5 | 740 | 140 | |
NM_020706.2 | NP_065757.1 | 4 - 5 | 740 | 140 | |
XM_005261017.2 | XP_005261074.1 | 4 - 5 | 721 | 140 | |
XM_006724035.2 | XP_006724098.1 | 4 - 5 | 721 | 140 | |
XM_006724036.2 | XP_006724099.1 | 4 - 5 | 721 | 140 | |
XM_017028415.1 | XP_016883904.1 | 4 - 5 | 701 | 140 | |
XM_017028416.1 | XP_016883905.1 | 4 - 5 | 586 | 140 | |
XM_017028417.1 | XP_016883906.1 | 4 - 5 | 721 | 140 | |
GenBank mRNA | AF023142.1 | - | 4 - 5 | 344 | 140 |
AK057840.1 | - | 4 - 5 | 583 | 140 | |
AK308406.1 | - | 3 - 4 | 545 | 140 | |
AL834304.1 | - | 4 - 5 | 520 | 140 | |
BC014921.1 | - | 4 - 5 | 717 | 140 | |
BC043353.1 | - | 4 - 5 | 697 | 140 | |
BC052286.1 | - | 4 - 5 | 571 | 140 | |
BC064990.1 | - | 4 - 5 | 571 | 140 |