Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
Chromosome Location
Chr.21: 31671000 - 31732118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145444.1 NP_001138916.1 13 - 14 2109 140
NM_001145445.1 NP_001138917.1 14 - 15 2154 140
NM_020706.2 NP_065757.1 14 - 15 2154 140
XM_005261017.2 XP_005261074.1 13 - 14 1958 140
XM_006724035.2 XP_006724098.1 14 - 15 2123 140
XM_006724036.2 XP_006724099.1 14 - 15 2123 140
XM_017028415.1 XP_016883904.1 14 - 15 2112 140
XM_017028416.1 XP_016883905.1 14 - 15 1997 140
XM_017028417.1 XP_016883906.1 13 - 14 1958 140
GenBank mRNA AB032998.1 - 8 - 9 1142 140
AK025748.1 - 9 - 10 1173 140
AK308406.1 - 13 - 14 1960 140
AL117417.1 - 9 - 10 1162 140
AL834304.1 - 14 - 15 1934 140
BC014921.1 - 14 - 15 2131 140
BC043353.1 - 14 - 15 2111 140
BC052286.1 - 14 - 15 1985 140
BC064990.1 - 14 - 15 1985 140

View Full Product Details