Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTHFSD
Entrez Gene ID
64779
Gene Name
methenyltetrahydrofolate synthetase domain containing
Gene Aliases
-
Chromosome Location
Chr.16: 86530176 - 86555235 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159377.1 NP_001152849.1 1 - 2 70 139
NM_001159378.1 NP_001152850.1 1 - 2 70 139
NR_027490.1 1 - 2 70 139
XM_011523284.2 XP_011521586.1 1 - 2 70 139
GenBank mRNA AK023060.1 - 1 - 2 45 139
BC112057.1 - 1 - 2 50 139
BC112059.1 - 1 - 2 50 139
BC143707.1 - 1 - 2 38 139

View Full Product Details