The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159377.1 | NP_001152849.1 | 4 - 5 | 404 | 128 |
NM_001159378.1 | NP_001152850.1 | 4 - 5 | 404 | 128 | |
NM_001159379.1 | NP_001152851.1 | 4 - 5 | 401 | 128 | |
NM_001159380.1 | NP_001152852.1 | 4 - 5 | 368 | 128 | |
NM_022764.2 | NP_073601.2 | 4 - 5 | 401 | 128 | |
NR_027490.1 | 4 - 5 | 464 | 128 | ||
XM_005256101.1 | XP_005256158.1 | 4 - 5 | 444 | 128 | |
XM_006721247.3 | XP_006721310.1 | 3 - 4 | 275 | 128 | |
XM_011523280.2 | XP_011521582.1 | 4 - 5 | 444 | 128 | |
XM_011523282.2 | XP_011521584.1 | 4 - 5 | 365 | 128 | |
XM_011523283.2 | XP_011521585.1 | 4 - 5 | 365 | 128 | |
XM_011523284.2 | XP_011521586.1 | 4 - 5 | 404 | 128 | |
XM_017023571.1 | XP_016879060.1 | 3 - 4 | 290 | 128 | |
XM_017023572.1 | XP_016879061.1 | 3 - 4 | 287 | 128 | |
GenBank mRNA | AK023060.1 | - | 4 - 5 | 379 | 128 |
AK023955.1 | - | 4 - 5 | 358 | 128 | |
AK297726.1 | - | 4 - 5 | 401 | 128 | |
BC013911.2 | - | 3 - 4 | 335 | 128 | |
BC112057.1 | - | 4 - 5 | 384 | 128 | |
BC112059.1 | - | 4 - 5 | 384 | 128 | |
BC143702.1 | - | 4 - 5 | 369 | 128 | |
BC143704.1 | - | 4 - 5 | 369 | 128 | |
BC143705.1 | - | 4 - 5 | 368 | 128 | |
BC143707.1 | - | 4 - 5 | 372 | 128 |