Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RMND5B
Entrez Gene ID
64777
Gene Name
required for meiotic nuclear division 5 homolog B
Gene Aliases
GID2, GID2B
Chromosome Location
Chr.5: 178130532 - 178148570 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288794.1 NP_001275723.1 11 - 12 1502 71
NM_001288795.1 NP_001275724.1 10 - 11 1351 71
NM_022762.4 NP_073599.2 10 - 11 1362 71
XM_005265969.3 XP_005266026.1 9 - 10 1182 71
XM_005265971.4 XP_005266028.1 10 - 11 1929 71
XM_005265972.3 XP_005266029.1 9 - 10 1309 71
XM_017009727.1 XP_016865216.1 9 - 10 1973 71
XM_017009728.1 XP_016865217.1 9 - 10 1640 71
GenBank mRNA AK025971.1 - 7 - 8 907 71
AK093481.1 - 10 - 11 1303 71
AK094065.1 - 7 - 8 1509 71
AK122692.1 - 5 - 6 2225 71
AK125337.1 - 11 - 12 1449 71
AL137381.1 - 4 - 5 475 71
AL713670.1 - 9 - 10 1225 71
AY359092.1 - 7 - 8 2220 71
BC009911.2 - 10 - 11 1296 71
CA440193.1 - 1 - 2 70
DQ494789.1 - 10 - 11 1296 71

View Full Product Details