The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012294.3 | NP_036426.3 | 16 - 17 | 1487 | 87 |
XM_005249914.2 | XP_005249971.1 | 16 - 17 | 1706 | 87 | |
XM_005249916.1 | XP_005249973.1 | 16 - 17 | 1650 | 87 | |
XM_011515652.2 | XP_011513954.1 | 8 - 9 | 3460 | 87 | |
XM_017012837.1 | XP_016868326.1 | 14 - 15 | 1391 | 87 | |
GenBank mRNA | AK295859.1 | - | 6 - 7 | 972 | 87 |
BC039203.1 | - | 6 - 7 | 798 | 87 | |
D87467.1 | - | 6 - 7 | 779 | 87 | |
KU178490.1 | - | 6 - 7 | 724 | 87 | |
KU178491.1 | - | 6 - 7 | 724 | 87 |