The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110556.1 | NP_001104026.1 | 20 - 21 | 3196 | 69 |
NM_001456.3 | NP_001447.2 | 20 - 21 | 3196 | 69 | |
XM_011531127.1 | XP_011529429.1 | 20 - 21 | 3191 | 69 | |
XM_011531128.1 | XP_011529430.1 | 20 - 21 | 3191 | 69 | |
XM_011531129.1 | XP_011529431.1 | 20 - 21 | 3191 | 69 | |
XM_011531130.1 | XP_011529432.1 | 20 - 21 | 3191 | 69 | |
XM_011531131.1 | XP_011529433.1 | 19 - 20 | 2990 | 69 | |
GenBank mRNA | AB191259.1 | - | 19 - 20 | 2919 | 69 |
AB191260.1 | - | 19 - 20 | 2924 | 69 | |
AB371574.1 | - | 19 - 20 | 2924 | 69 | |
AB371575.1 | - | 19 - 20 | 2919 | 69 | |
AB371576.1 | - | 19 - 20 | 2924 | 69 | |
AB371577.1 | - | 19 - 20 | 2919 | 69 | |
AB371578.1 | - | 19 - 20 | 2919 | 69 | |
AB371579.1 | - | 19 - 20 | 2919 | 69 | |
AB593010.1 | - | 19 - 20 | 3059 | 69 | |
AK090427.2 | - | 19 - 20 | 2985 | 69 | |
GU727643.1 | - | 20 - 21 | 3196 | 69 | |
X53416.1 | - | 20 - 21 | 3119 | 69 |