The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110556.1 | NP_001104026.1 | 27 - 28 | 4846 | 80 |
NM_001456.3 | NP_001447.2 | 27 - 28 | 4846 | 80 | |
XM_011531127.1 | XP_011529429.1 | 27 - 28 | 4841 | 80 | |
XM_011531128.1 | XP_011529430.1 | 27 - 28 | 4841 | 80 | |
XM_011531129.1 | XP_011529431.1 | 27 - 28 | 4841 | 80 | |
XM_011531130.1 | XP_011529432.1 | 27 - 28 | 4841 | 80 | |
XM_011531131.1 | XP_011529433.1 | 26 - 27 | 4640 | 80 | |
GenBank mRNA | AB191259.1 | - | 26 - 27 | 4569 | 80 |
AB191260.1 | - | 26 - 27 | 4574 | 80 | |
AB371574.1 | - | 26 - 27 | 4574 | 80 | |
AB371575.1 | - | 26 - 27 | 4569 | 80 | |
AB371576.1 | - | 26 - 27 | 4574 | 80 | |
AB371577.1 | - | 26 - 27 | 4569 | 80 | |
AB371578.1 | - | 26 - 27 | 4569 | 80 | |
AB371579.1 | - | 26 - 27 | 4569 | 80 | |
AB593010.1 | - | 26 - 27 | 4709 | 80 | |
AK074048.1 | - | 6 - 7 | 1046 | 80 | |
AK090427.2 | - | 26 - 27 | 4635 | 80 | |
AL157419.1 | - | 4 - 5 | 610 | 80 | |
GU727643.1 | - | 27 - 28 | 4846 | 80 | |
X53416.1 | - | 27 - 28 | 4769 | 80 |