Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
Chromosome Location
Chr.X: 154348529 - 154374638 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110556.1 NP_001104026.1 32 - 33 5562 89
NM_001456.3 NP_001447.2 31 - 32 5538 89
XM_011531129.1 XP_011529431.1 32 - 33 5557 89
XM_011531130.1 XP_011529432.1 31 - 32 5533 89
XM_011531131.1 XP_011529433.1 31 - 32 5356 89
GenBank mRNA AB191259.1 - 30 - 31 5261 89
AB191260.1 - 31 - 32 5290 89
AB371574.1 - 31 - 32 5290 89
AB371575.1 - 30 - 31 5261 89
AB371576.1 - 31 - 32 5290 89
AB371577.1 - 31 - 32 5285 89
AB371578.1 - 30 - 31 5261 89
AB371579.1 - 30 - 31 5261 89
AB593010.1 - 31 - 32 5425 89
AK074048.1 - 10 - 11 1738 89
AK090427.2 - 30 - 31 5327 89
AK300165.1 - 5 - 6 598 89
AK304255.1 - 4 - 5 573 89
AL050396.1 - 4 - 5 493 89
AL157419.1 - 8 - 9 1302 89
BC014654.1 - 2 - 3 204 89
BC067111.1 - 2 - 3 323 89
GU727643.1 - 32 - 33 5562 89
X53416.1 - 32 - 33 5485 89

View Full Product Details