Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TOMM40L
- Entrez Gene ID
-
84134
- Gene Name
- translocase of outer mitochondrial membrane 40 like
- Gene Aliases
-
TOMM40B
- Chromosome Location
-
Chr.1: 161225939 - 161230746 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK297831.1 |
- |
2 - 3 |
307 |
58 |
View Full Product Details