The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320818.1 | NP_001307747.1 | 2 - 3 | 433 | 113 |
NM_001320819.1 | NP_001307748.1 | 3 - 4 | 627 | 113 | |
NM_001320820.1 | NP_001307749.1 | 3 - 4 | 824 | 113 | |
NM_025147.4 | NP_079423.1 | 3 - 4 | 583 | 113 | |
GenBank mRNA | AK023510.1 | - | 3 - 4 | 452 | 113 |
AK294093.1 | - | 3 - 4 | 824 | 113 | |
AK308977.1 | - | 3 - 4 | 421 | 113 | |
BC022453.1 | - | 3 - 4 | 500 | 113 | |
BU195968.1 | - | 2 - 3 | 367 | 113 | |
DC328985.1 | - | 3 - 4 | 421 | 113 |