Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMBOX1
Entrez Gene ID
79618
Gene Name
homeobox containing 1
Gene Aliases
HNF1LA, HOT1, PBHNF
Chromosome Location
Chr.8: 28890370 - 29064939 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135726.2 NP_001129198.1 2 - 3 388 94
NM_001324382.1 NP_001311311.1 2 - 3 248 94
NM_001324383.1 NP_001311312.1 2 - 3 388 94
NM_001324384.1 NP_001311313.1 2 - 3 391 94
NM_001324385.1 NP_001311314.1 2 - 3 388 94
NM_001324386.1 NP_001311315.1 2 - 3 388 94
NM_001324387.1 NP_001311316.1 2 - 3 388 94
NM_001324388.1 NP_001311317.1 2 - 3 248 94
NM_001324389.1 NP_001311318.1 2 - 3 388 94
NM_001324390.1 NP_001311319.1 2 - 3 391 94
NM_001324391.1 NP_001311320.1 2 - 3 388 94
NM_001324392.1 NP_001311321.1 2 - 3 391 94
NM_001324393.1 NP_001311322.1 3 - 4 504 94
NM_001324394.1 NP_001311323.1 2 - 3 388 94
NM_001324395.1 NP_001311324.1 2 - 3 388 94
NM_024567.4 NP_078843.2 3 - 4 422 94
NR_136757.1 2 - 3 388 94
NR_136758.1 2 - 3 388 94
XM_005273635.1 XP_005273692.1 2 - 3 364 94
XM_005273641.4 XP_005273698.1 2 - 3 364 94
XM_006716398.2 XP_006716461.1 2 - 3 364 94
XM_011544642.2 XP_011542944.1 2 - 3 364 94
XM_017013823.1 XP_016869312.1 2 - 3 396 94
XM_017013824.1 XP_016869313.1 2 - 3 186 94
XM_017013825.1 XP_016869314.1 2 - 3 364 94
GenBank mRNA AK290683.1 - 2 - 3 364 94
AK310959.1 - 2 - 3 354 94
AY522342.1 - 2 - 3 133 94
BC069242.1 - 3 - 4 422 94
DC341559.1 - 3 - 4 255 94
DC358620.1 - 2 - 3 208 94
DC395744.1 - 2 - 3 249 94
DQ269478.1 - 2 - 3 133 94

View Full Product Details