Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HMBOX1
- Entrez Gene ID
-
79618
- Gene Name
- homeobox containing 1
- Gene Aliases
-
HNF1LA, HOT1, PBHNF
- Chromosome Location
-
Chr.8: 28890370 - 29064939 on Build GRCh38
View Full Product Details