The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144063.1 | NP_001137535.1 | 18 - 19 | 2210 | 137 |
NM_020896.3 | NP_065947.1 | 19 - 20 | 2414 | 137 | |
NM_145638.2 | NP_663613.1 | 18 - 19 | 2203 | 137 | |
XM_011519873.2 | XP_011518175.1 | 19 - 20 | 2753 | 137 | |
XM_017017163.1 | XP_016872652.1 | 19 - 20 | 2761 | 137 | |
XM_017017164.1 | XP_016872653.1 | 14 - 15 | 2332 | 137 | |
GenBank mRNA | AB074006.1 | - | 19 - 20 | 2355 | 137 |
AF392453.1 | - | 19 - 20 | 2327 | 137 | |
AF410430.1 | - | 18 - 19 | 2272 | 137 | |
AK055617.1 | - | 14 - 15 | 1748 | 137 | |
AK056510.1 | - | 19 - 20 | 2372 | 137 | |
AK124919.1 | - | 5 - 6 | 1180 | 137 | |
AK293000.1 | - | 18 - 19 | 2159 | 137 | |
AK297475.1 | - | 15 - 16 | 1509 | 137 | |
AK301003.1 | - | 17 - 18 | 2050 | 137 | |
AL136918.1 | - | 19 - 20 | 2327 | 137 | |
BC017505.1 | - | 10 - 11 | 1070 | 137 | |
BC032646.1 | - | 18 - 19 | 2124 | 137 | |
BC039579.1 | - | 19 - 20 | 2325 | 137 |