Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAMD4
Entrez Gene ID
23151
Gene Name
GRAM domain containing 4
Gene Aliases
DIP, dA59H18.1, dJ439F8.1
Chromosome Location
Chr.22: 46576017 - 46682755 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015124.4 NP_055939.1 10 - 11 1076 70
XM_005261396.3 XP_005261453.1 10 - 11 1012 70
XM_006724169.1 XP_006724232.1 10 - 11 974 70
XM_006724170.3 XP_006724233.1 10 - 11 993 70
XM_011530011.1 XP_011528313.1 9 - 10 912 70
XM_011530012.1 XP_011528314.1 9 - 10 912 70
XM_011530013.2 XP_011528315.2 10 - 11 1021 70
XM_011530014.2 XP_011528316.1 9 - 10 912 70
XM_017028670.1 XP_016884159.1 10 - 11 1034 70
GenBank mRNA AB018310.2 - 10 - 11 1076 70
BC129837.1 - 9 - 10 897 70
CR456470.1 - 9 - 10 902 70

View Full Product Details