The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005915.5 | NP_005906.2 | 13 - 14 | 2022 | 140 |
GenBank mRNA | AK299236.1 | - | 12 - 13 | 1671 | 140 |
AK312575.1 | - | 13 - 14 | 1969 | 140 | |
BC020268.1 | - | 13 - 14 | 1950 | 140 | |
BC032374.1 | - | 13 - 14 | 1965 | 140 | |
D84557.1 | - | 13 - 14 | 1971 | 140 | |
U46838.1 | - | 13 - 14 | 1959 | 140 |