The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005915.5 | NP_005906.2 | 4 - 5 | 723 | 89 |
GenBank mRNA | AK299236.1 | - | 3 - 4 | 372 | 89 |
AK303455.1 | - | 3 - 4 | 467 | 89 | |
AK312575.1 | - | 4 - 5 | 670 | 89 | |
BC020268.1 | - | 4 - 5 | 651 | 89 | |
BC032374.1 | - | 4 - 5 | 666 | 89 | |
D84557.1 | - | 4 - 5 | 672 | 89 | |
U46838.1 | - | 4 - 5 | 660 | 89 |