The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080473.4 | NP_536721.1 | 6 - 7 | 1103 | 86 |
XM_006723699.3 | XP_006723762.1 | 6 - 7 | 1123 | 86 | |
GenBank mRNA | BC047790.1 | - | 6 - 7 | 1103 | 86 |
BC117356.1 | - | 5 - 6 | 1066 | 86 | |
BC117358.1 | - | 5 - 6 | 1066 | 86 |