Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPSS1
Entrez Gene ID
9061
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 1
Gene Aliases
ATPSK1, PAPSS, SK1
Chromosome Location
Chr.4: 107613666 - 107720263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005443.4 NP_005434.4 10 - 11 1593 79
XM_011532400.1 XP_011530702.1 10 - 11 1573 79
XM_011532401.1 XP_011530703.1 10 - 11 1659 79
GenBank mRNA AF033026.1 - 10 - 11 1535 79
AF105227.1 - 10 - 11 1563 79
AK292774.1 - 10 - 11 1543 79
BC011392.1 - 10 - 11 1466 79
BC032513.1 - 10 - 11 1562 79
BC040940.1 - 10 - 11 1521 79
BC050627.1 - 10 - 11 1529 79
CR457028.1 - 10 - 11 1509 79
Y10387.1 - 10 - 11 1545 79

View Full Product Details