The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014448.3 | NP_055263.2 | 9 - 10 | 1633 | 72 |
XM_017001049.1 | XP_016856538.1 | 9 - 10 | 2234 | 72 | |
XM_017001050.1 | XP_016856539.1 | 9 - 10 | 1439 | 72 | |
XM_017001051.1 | XP_016856540.1 | 9 - 10 | 1644 | 72 | |
GenBank mRNA | AK091910.1 | - | 3 - 4 | 672 | 72 |
AK095984.1 | - | 9 - 10 | 1540 | 72 | |
AK296150.1 | - | 4 - 5 | 531 | 72 | |
BC002681.1 | - | 8 - 9 | 920 | 72 | |
BC051838.1 | - | 7 - 8 | 940 | 72 | |
BT007270.1 | - | 4 - 5 | 521 | 72 | |
D89016.1 | - | 6 - 7 | 948 | 72 |