The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000096.3 | NP_000087.1 | 11 - 12 | 2328 | 82 |
XM_006713499.2 | XP_006713562.1 | 11 - 12 | 2333 | 82 | |
XM_006713500.3 | XP_006713563.1 | 11 - 12 | 2333 | 82 | |
XM_006713501.2 | XP_006713564.1 | 11 - 12 | 2333 | 82 | |
XM_011512435.1 | XP_011510737.1 | 11 - 12 | 2333 | 82 | |
XM_017005734.1 | XP_016861223.1 | 11 - 12 | 2333 | 82 | |
XM_017005735.1 | XP_016861224.1 | 11 - 12 | 2333 | 82 | |
GenBank mRNA | AK095290.1 | - | 10 - 11 | 1617 | 82 |
AK291219.1 | - | 11 - 12 | 2110 | 82 | |
BC094709.1 | - | 11 - 12 | 2112 | 82 | |
BC142714.1 | - | 11 - 12 | 2084 | 82 | |
BC146663.1 | - | 11 - 12 | 2084 | 82 | |
BC146801.1 | - | 11 - 12 | 2108 | 82 | |
M13536.1 | - | 8 - 9 | 1425 | 82 | |
M13699.1 | - | 11 - 12 | 2075 | 82 |