The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000096.3 | NP_000087.1 | 2 - 3 | 649 | 91 |
NR_046371.1 | 2 - 3 | 649 | 91 | ||
XM_006713499.2 | XP_006713562.1 | 2 - 3 | 654 | 91 | |
XM_006713500.3 | XP_006713563.1 | 2 - 3 | 654 | 91 | |
XM_006713501.2 | XP_006713564.1 | 2 - 3 | 654 | 91 | |
XM_011512435.1 | XP_011510737.1 | 2 - 3 | 654 | 91 | |
XM_017005734.1 | XP_016861223.1 | 2 - 3 | 654 | 91 | |
XM_017005735.1 | XP_016861224.1 | 2 - 3 | 654 | 91 | |
GenBank mRNA | AK291219.1 | - | 2 - 3 | 431 | 91 |
AK299272.1 | - | 2 - 3 | 435 | 91 | |
BC070304.1 | - | 2 - 3 | 602 | 91 | |
BC094709.1 | - | 2 - 3 | 433 | 91 | |
BC142714.1 | - | 2 - 3 | 405 | 91 | |
BC146663.1 | - | 2 - 3 | 405 | 91 | |
BC146801.1 | - | 2 - 3 | 429 | 91 | |
CA391602.1 | - | 2 - 3 | 646 | 91 | |
M13699.1 | - | 2 - 3 | 396 | 91 |