The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004501.3 | NP_004492.2 | 11 - 12 | 2324 | 96 |
NM_031844.2 | NP_114032.2 | 11 - 12 | 2381 | 96 | |
XM_017001115.1 | XP_016856604.1 | 11 - 12 | 2576 | 96 | |
XM_017001116.1 | XP_016856605.1 | 11 - 12 | 2381 | 96 | |
XM_017001117.1 | XP_016856606.1 | 11 - 12 | 2370 | 96 | |
GenBank mRNA | AF068846.1 | - | 11 - 12 | 2377 | 96 |
AF461014.1 | - | 8 - 9 | 1902 | 96 | |
AK126868.1 | - | 12 - 13 | 2122 | 96 | |
AK130669.1 | - | 10 - 11 | 1430 | 96 | |
AK297119.1 | - | 12 - 13 | 2215 | 96 | |
BC003367.1 | - | 11 - 12 | 2255 | 96 | |
BC003621.2 | - | 11 - 12 | 2257 | 96 | |
BC007950.2 | - | 11 - 12 | 1805 | 96 | |
BC015782.1 | - | 11 - 12 | 1858 | 96 | |
BC024767.1 | - | 11 - 12 | 2257 | 96 | |
BC034925.1 | - | 8 - 9 | 1248 | 96 | |
X65488.1 | - | 11 - 12 | 2147 | 96 |