Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGO2
Entrez Gene ID
151246
Gene Name
shugoshin 2
Gene Aliases
SGOL2, TRIPIN
Chromosome Location
Chr.2: 200510198 - 200584095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160033.1 NP_001153505.1 4 - 5 493 145
NM_001160046.1 NP_001153518.1 4 - 5 493 145
NM_152524.5 NP_689737.4 4 - 5 493 145
XM_005246344.3 XP_005246401.2 5 - 6 1624 145
XM_005246345.3 XP_005246402.1 5 - 6 1697 145
XM_011510734.2 XP_011509036.1 4 - 5 412 145
XM_017003452.1 XP_016858941.1 6 - 7 1750 145
XM_017003453.1 XP_016858942.1 7 - 8 747 145
XM_017003454.1 XP_016858943.1 6 - 7 1759 145
GenBank mRNA AK095291.1 - 4 - 5 478 145
AK302457.1 - 4 - 5 432 145
AY094614.1 - 4 - 5 401 145
BC035764.1 - 4 - 5 446 145
BC048349.1 - 4 - 5 468 145
BC092412.1 - 4 - 5 418 145
BX647433.1 - 4 - 5 438 145

View Full Product Details