The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032301.2 | NP_115677.2 | 5 - 6 | 947 | 68 |
XM_005260096.4 | XP_005260153.1 | 4 - 5 | 1066 | 68 | |
GenBank mRNA | AK123037.1 | - | 5 - 6 | 946 | 68 |
AK302744.1 | - | 3 - 4 | 672 | 68 | |
BC004290.1 | - | 5 - 6 | 924 | 68 |