The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015523.3 | NP_056338.2 | 1 - 2 | 285 | 78 |
GenBank mRNA | AF151872.1 | - | 1 - 2 | 236 | 78 |
AK223200.1 | - | 1 - 2 | 200 | 78 | |
AK312042.1 | - | 1 - 2 | 214 | 78 | |
AL110239.1 | - | 1 - 2 | 219 | 78 | |
BC003502.1 | - | 1 - 2 | 187 | 78 | |
BC105024.1 | - | 1 - 2 | 206 | 78 | |
BC105026.1 | - | 1 - 2 | 206 | 78 | |
BC107887.1 | - | - | 195 | 78 | |
BC143701.1 | - | 1 - 2 | 209 | 78 | |
CR533500.1 | - | 1 - 2 | 142 | 78 | |
DB471224.1 | - | 1 - 2 | 286 | 78 |