The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015523.3 | NP_056338.2 | 6 - 7 | 725 | 107 |
GenBank mRNA | AF151872.1 | - | 6 - 7 | 676 | 107 |
AK023498.1 | - | 2 - 3 | 1022 | 107 | |
AK223200.1 | - | 6 - 7 | 640 | 107 | |
AK312042.1 | - | 6 - 7 | 654 | 107 | |
AL110239.1 | - | 6 - 7 | 659 | 107 | |
BC105024.1 | - | 6 - 7 | 646 | 107 | |
BC105026.1 | - | 6 - 7 | 646 | 107 | |
BC107887.1 | - | - | 635 | 107 | |
BC143701.1 | - | 6 - 7 | 649 | 107 | |
CR533500.1 | - | 6 - 7 | 582 | 107 |