The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017546.4 | NP_060016.3 | 3 - 4 | 915 | 70 |
GenBank mRNA | AF103798.1 | - | 3 - 4 | 906 | 70 |
AK024221.1 | - | 2 - 3 | 332 | 70 | |
BC018664.2 | - | 3 - 4 | 824 | 70 | |
BC064421.1 | - | 3 - 4 | 882 | 70 |