The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129887.1 | NP_001123359.1 | 13 - 14 | 1724 | 76 |
NM_014377.1 | NP_055192.1 | 15 - 16 | 1883 | 76 | |
XM_005250269.2 | XP_005250326.1 | 16 - 17 | 1832 | 76 | |
XM_011516028.2 | XP_011514330.1 | 15 - 16 | 2283 | 76 | |
XM_011516029.2 | XP_011514331.1 | 16 - 17 | 1744 | 76 | |
XM_011516030.2 | XP_011514332.1 | 11 - 12 | 1084 | 76 | |
XM_017011968.1 | XP_016867457.1 | 16 - 17 | 1919 | 76 | |
GenBank mRNA | AK126382.1 | - | 1 - 2 | 1585 | 76 |
AL832847.1 | - | 14 - 15 | 1667 | 76 | |
BC000859.1 | - | 7 - 8 | 727 | 76 | |
BC139751.1 | - | 15 - 16 | 1631 | 76 | |
X98260.1 | - | 13 - 14 | 1517 | 76 |