Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOBOX
Entrez Gene ID
135935
Gene Name
NOBOX oogenesis homeobox
Gene Aliases
OG-2, OG2, OG2X, POF5, TCAG_12042
Chromosome Location
Chr.7: 144396805 - 144410438 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080413.3 NP_001073882.3 5 - 6 1047 74

View Full Product Details