Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8orf37
Entrez Gene ID
157657
Gene Name
chromosome 8 open reading frame 37
Gene Aliases
BBS21, CORD16, RP64, smalltalk
Chromosome Location
Chr.8: 95244913 - 95270602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177965.3 NP_808880.1 3 - 4 358 74
XM_005250799.3 XP_005250856.2 3 - 4 1726 74
GenBank mRNA BC036855.2 - 3 - 4 323 74
DA473498.1 - 3 - 4 358 74
DB205618.1 - 3 - 4 323 74

View Full Product Details