The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177965.3 | NP_808880.1 | 4 - 5 | 413 | 72 |
GenBank mRNA | BC036855.2 | - | 4 - 5 | 378 | 72 |
DA473498.1 | - | 4 - 5 | 413 | 72 | |
DB205618.1 | - | 4 - 5 | 378 | 72 |