The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197259.1 | NP_001184188.1 | 17 - 18 | 1954 | 66 |
NM_012381.3 | NP_036513.2 | 18 - 19 | 2052 | 66 | |
NM_181837.2 | NP_862820.1 | 18 - 19 | 2055 | 66 | |
XM_005248704.1 | XP_005248761.1 | 18 - 19 | 2001 | 66 | |
XM_005248705.1 | XP_005248762.1 | 19 - 20 | 2059 | 66 | |
XM_005248706.1 | XP_005248763.1 | 19 - 20 | 2058 | 66 | |
XM_011535651.1 | XP_011533953.1 | 17 - 18 | 1887 | 66 | |
XM_017010632.1 | XP_016866121.1 | 18 - 19 | 2001 | 66 | |
XM_017010633.1 | XP_016866122.1 | 18 - 19 | 1998 | 66 | |
XM_017010634.1 | XP_016866123.1 | 18 - 19 | 1998 | 66 | |
XM_017010635.1 | XP_016866124.1 | 17 - 18 | 1884 | 66 | |
XM_017010636.1 | XP_016866125.1 | 19 - 20 | 2056 | 66 | |
XM_017010637.1 | XP_016866126.1 | 19 - 20 | 2055 | 66 | |
XM_017010638.1 | XP_016866127.1 | 19 - 20 | 2056 | 66 | |
XM_017010639.1 | XP_016866128.1 | 19 - 20 | 2055 | 66 | |
XM_017010640.1 | XP_016866129.1 | 18 - 19 | 1941 | 66 | |
GenBank mRNA | AF093535.1 | - | 18 - 19 | 1976 | 66 |
AF125507.1 | - | 18 - 19 | 1985 | 66 | |
AF135044.1 | - | 18 - 19 | 1950 | 66 | |
AK222795.1 | - | 18 - 19 | 1978 | 66 | |
AK303182.1 | - | 17 - 18 | 1773 | 66 | |
AK303195.1 | - | 17 - 18 | 1878 | 66 | |
AK315911.1 | - | 18 - 19 | 1939 | 66 | |
AL080116.1 | - | 18 - 19 | 1994 | 66 | |
BC035494.2 | - | 18 - 19 | 1967 | 66 | |
BC047689.1 | - | 18 - 19 | 1971 | 66 | |
BM971022.1 | - | 2 - 3 | 66 | ||
U50950.1 | - | 14 - 15 | 1618 | 66 |