Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFAP1L2
Entrez Gene ID
84632
Gene Name
actin filament associated protein 1 like 2
Gene Aliases
CTB-1144G6.4, KIAA1914, XB130
Chromosome Location
Chr.10: 114280730 - 114405143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001936.2 NP_001001936.1 13 - 14 1861 116
NM_001287824.1 NP_001274753.1 14 - 15 2020 116
NM_032550.3 NP_115939.1 13 - 14 1861 116
XM_005270233.4 XP_005270290.1 14 - 15 1809 116
XM_005270237.4 XP_005270294.1 13 - 14 1585 116
XM_005270241.4 XP_005270298.1 14 - 15 1820 116
XM_005270242.4 XP_005270299.1 11 - 12 1407 116
XM_011540286.2 XP_011538588.1 13 - 14 1631 116
XM_011540289.1 XP_011538591.1 13 - 14 1631 116
XM_011540290.1 XP_011538592.1 13 - 14 1631 116
XM_017016792.1 XP_016872281.1 14 - 15 1715 116
XM_017016793.1 XP_016872282.1 14 - 15 1715 116
XM_017016794.1 XP_016872283.1 14 - 15 1715 116
XM_017016795.1 XP_016872284.1 15 - 16 1893 116
XM_017016796.1 XP_016872285.1 14 - 15 1667 116
XM_017016797.1 XP_016872286.1 14 - 15 1667 116
XM_017016798.1 XP_016872287.1 14 - 15 1667 116
XM_017016799.1 XP_016872288.1 14 - 15 1715 116
XM_017016800.1 XP_016872289.1 14 - 15 1715 116
XM_017016801.1 XP_016872290.1 14 - 15 1715 116
XM_017016802.1 XP_016872291.1 14 - 15 1715 116
XM_017016803.1 XP_016872292.1 14 - 15 1715 116
XM_017016804.1 XP_016872293.1 13 - 14 1585 116
XM_017016805.1 XP_016872294.1 14 - 15 1667 116
XM_017016806.1 XP_016872295.1 14 - 15 1667 116
XM_017016807.1 XP_016872296.1 13 - 14 1631 116
XM_017016808.1 XP_016872297.1 14 - 15 1667 116
XM_017016809.1 XP_016872298.1 14 - 15 1667 116
XM_017016810.1 XP_016872299.1 14 - 15 1809 116
XM_017016811.1 XP_016872300.1 14 - 15 1809 116
XM_017016812.1 XP_016872301.1 14 - 15 1809 116
XM_017016813.1 XP_016872302.1 14 - 15 1809 116
XM_017016814.1 XP_016872303.1 15 - 16 1904 116
XM_017016816.1 XP_016872305.1 13 - 14 1585 116
XM_017016817.1 XP_016872306.1 13 - 14 1585 116
XM_017016820.1 XP_016872309.1 15 - 16 1904 116
XM_017016821.1 XP_016872310.1 15 - 16 1904 116
XM_017016825.1 XP_016872314.1 14 - 15 1820 116
XM_017016826.1 XP_016872315.1 14 - 15 1820 116
XM_017016829.1 XP_016872318.1 12 - 13 1489 116
XM_017016831.1 XP_016872320.1 12 - 13 1486 116
XM_017016832.1 XP_016872321.1 11 - 12 1449 116
GenBank mRNA AB067501.1 - 1 - 2 660 116
AF474151.1 - 13 - 14 1587 116
AK123108.1 - 14 - 15 1631 116
AK291712.1 - 13 - 14 1585 116
AK294969.1 - 14 - 15 1746 116
AK295538.1 - 3 - 4 482 116
BC024314.1 - 13 - 14 1839 116
BC033212.1 - 13 - 14 1568 116

View Full Product Details